siRNA Details
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080492.1 | NP_001073961.1 | 5 | 702 | ||
NM_001305173.1 | NP_001292102.1 | 5 | 699 | |||
NM_001305174.1 | NP_001292103.1 | 4 | 524 | |||
GenBank mRNA | AK058068.1 | - | 5 | 644 | ||
AK302175.1 | - | 4 | 454 | |||
BC057843.1 | - | 5 | 691 | 5296090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
