siRNA Details
Gene Symbol: ZNF500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303450.1 | NP_001290379.1 | 7 | 2215 | ||
NM_021646.3 | NP_067678.1 | 6 | 3078 | |||
XM_005255243.3 | XP_005255300.1 | 5 | 3044 | |||
XM_011522454.2 | XP_011520756.1 | 5 | 2833 | |||
XM_017023121.1 | XP_016878610.1 | 5 | 2571 | |||
GenBank mRNA | AB011129.1 | - | 5 | 2818 | ||
AF007129.1 | - | 1 | 1333 | |||
AK297993.1 | - | 7 | 2214 | |||
BC004125.2 | - | 1 | 2019 | |||
BC033151.1 | - | 1 | 2148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
