siRNA Details
Gene Symbol: KNOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012991.2 | NP_001013009.2 | 4 | 1715 | ||
XM_005255317.4 | XP_005255374.1 | 6 | 1910 | |||
XM_006721046.3 | XP_006721109.1 | 5 | 2073 | |||
XM_011545855.1 | XP_011544157.1 | 5 | 1959 | |||
XM_017023231.1 | XP_016878720.1 | 5 | 1851 | |||
XM_017023232.1 | XP_016878721.1 | 4 | 1996 | |||
GenBank mRNA | BC117562.1 | - | 4 | 1715 | 7971177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
