siRNA Details
Gene Symbol: PPP1R37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019121.1 | NP_061994.1 | 5 | 631 | ||
XM_006723159.2 | XP_006723222.1 | 5 | 631 | |||
GenBank mRNA | BC035704.1 | - | 5 | 853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
