siRNA Details
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 5, 6 | 1065 | |||
NR_104095.1 | 4, 5 | 745 | ||||
NR_104096.1 | 4, 5 | 727 | ||||
NR_104097.1 | 4, 5 | 727 | ||||
NR_136268.1 | 5, 6 | 1065 | ||||
GenBank mRNA | AK024467.1 | - | 2, 3 | 918 | ||
BC033195.1 | - | 1, 2 | 223 | |||
BC171827.1 | - | 4, 5 | 745 | |||
BC171832.1 | - | 4, 5 | 727 | |||
BC171834.1 | - | 4, 5 | 999 | |||
BC171838.1 | - | 4, 5 | 727 | |||
BC171839.1 | - | 3, 4 | 940 | |||
DQ224421.1 | - | 5, 6 | 966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
