siRNA Details
Gene Symbol: CACTIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080543.1 | NP_001074012.1 | 10 | 3390 | ||
NM_021231.1 | NP_067054.1 | 11 | 2458 | |||
GenBank mRNA | AF052087.1 | - | 1 | 860 | ||
AF155102.1 | - | 3 | 806 | |||
AK299963.1 | - | 1724 | ||||
AY917150.1 | - | 12 | 2596 | |||
BC004262.1 | - | 2 | 1612 | |||
BC019848.1 | - | 4 | 1002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
