siRNA Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300974.1 | NP_001287903.1 | 4 | 885 | ||
NM_014209.3 | NP_055024.2 | 5 | 982 | |||
XM_005258652.2 | XP_005258709.1 | 4 | 1046 | |||
XM_011526624.2 | XP_011524926.1 | 4 | 1463 | |||
XM_017026472.1 | XP_016881961.1 | 3 | 965 | |||
GenBank mRNA | AK097439.1 | - | 4 | 1046 | ||
BC040908.1 | - | 4 | 829 | |||
BC140746.1 | - | 5 | 672 | 9021614 | ||
BC144448.1 | - | 5 | 670 | 9052974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
