siRNA Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300974.1 | NP_001287903.1 | 4, 5 | 1059 | ||
NM_014209.3 | NP_055024.2 | 5, 6 | 1156 | |||
XM_005258652.2 | XP_005258709.1 | 4, 5 | 1220 | |||
XM_011526624.2 | XP_011524926.1 | 4, 5 | 1637 | |||
XM_017026472.1 | XP_016881961.1 | 3, 4 | 1139 | |||
GenBank mRNA | AF000671.1 | - | 1, 2 | 105 | ||
AK097439.1 | - | 4, 5 | 1220 | |||
BC040908.1 | - | 4, 5 | 1003 | |||
BC107123.1 | - | 6, 7 | 602 | |||
BC140746.1 | - | 5, 6 | 846 | 9021614 | ||
BC144448.1 | - | 5, 6 | 844 | 9052974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
