siRNA Details
Gene Symbol: ETV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300974.1 | NP_001287903.1 | 4, 5 | 1061 | ||
NM_014209.3 | NP_055024.2 | 5, 6 | 1158 | |||
XM_005258652.2 | XP_005258709.1 | 4, 5 | 1222 | |||
XM_011526624.2 | XP_011524926.1 | 4, 5 | 1639 | |||
XM_017026472.1 | XP_016881961.1 | 3, 4 | 1141 | |||
GenBank mRNA | AF000671.1 | - | 1, 2 | 107 | ||
AK097439.1 | - | 4, 5 | 1222 | |||
BC040908.1 | - | 4, 5 | 1005 | |||
BC107123.1 | - | 6, 7 | 604 | |||
BC140746.1 | - | 5, 6 | 848 | 9021614 | ||
BC144448.1 | - | 5, 6 | 846 | 9052974 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007959, L10427, AB221612.
Gene Ontology Categories:
