siRNA Details
Gene Symbol: STEAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207342.2 | NP_997225.1 | 5 | 1155 | ||
GenBank mRNA | BC066301.1 | - | 5 | 1052 | 5274319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
