siRNA Details
Gene Symbol: LFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040167.1 | NP_001035257.1 | 7 | 1027 | ||
NM_001040168.1 | NP_001035258.1 | 7 | 1027 | |||
NM_001166355.1 | NP_001159827.1 | 8 | 923 | |||
NM_002304.2 | NP_002295.1 | 8 | 711 | |||
GenBank mRNA | AF193612.1 | - | 7 | 702 | ||
AK096284.1 | - | 8 | 923 | |||
BC014851.1 | - | 8 | 711 | 4453156 | ||
BM854210.1 | - | 2 | 125 | |||
U94354.1 | - | 7 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
