siRNA Details
Gene Symbol: LFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040167.1 | NP_001035257.1 | 6 | 861 | ||
NM_001040168.1 | NP_001035258.1 | 6 | 861 | |||
NM_001166355.1 | NP_001159827.1 | 7 | 757 | |||
NM_002304.2 | NP_002295.1 | 7 | 545 | |||
GenBank mRNA | AF193612.1 | - | 6 | 536 | ||
AK096284.1 | - | 7 | 757 | |||
BC014851.1 | - | 7 | 545 | 4453156 | ||
U94354.1 | - | 6 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
