siRNA Details
Gene Symbol: NOBOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080413.3 | NP_001073882.3 | 4 | 806 | ||
XM_017011742.1 | XP_016867231.1 | 4 | 1017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
