siRNA Details
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138426.3 | NP_612435.1 | 8 | 2523 | ||
GenBank mRNA | AK055741.1 | - | 1 | 1924 | ||
AK126731.1 | - | 2 | 1674 | |||
AL832860.1 | - | 2 | 635 | |||
AL833694.1 | - | 4 | 1018 | |||
BC011254.1 | - | 2 | 731 | |||
BC050291.1 | - | 8 | 2473 | 5272003 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286728, NM_001286729, NM_133236, AK088359, BC050802, and others.
