siRNA Details
Gene Symbol: ZNF862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099220.1 | NP_001092690.1 | 4 | 978 | ||
XM_006716085.3 | XP_006716148.1 | 5 | 999 | |||
XM_011516463.1 | XP_011514765.1 | 4 | 1859 | |||
XM_011516464.1 | XP_011514766.1 | 4 | 2248 | |||
XM_017012519.1 | XP_016868008.1 | 4 | 988 | |||
GenBank mRNA | AB011115.1 | - | 2 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
