siRNA Details
Gene Symbol: MEGF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080497.2 | NP_001073966.2 | 3 | 993 | ||
GenBank mRNA | AB011542.1 | - | 2 | 185 | ||
AK295380.1 | - | 3 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
