siRNA Details
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 14 | 3285 | ||
XM_005270294.4 | XP_005270351.1 | 15 | 3418 | |||
XM_005270295.4 | XP_005270352.1 | 14 | 3372 | |||
XM_005270297.3 | XP_005270354.1 | 9 | 4649 | |||
XM_005270298.4 | XP_005270355.1 | 9 | 3452 | |||
XM_006718079.3 | XP_006718142.1 | 16 | 3515 | |||
XM_006718080.3 | XP_006718143.1 | 10 | 3546 | |||
XM_011540385.2 | XP_011538687.1 | 15 | 3469 | |||
XM_011540386.2 | XP_011538688.1 | 10 | 4745 | |||
XM_011540387.2 | XP_011538689.1 | 6 | 6391 | |||
XM_017016969.1 | XP_016872458.1 | 15 | 3436 | |||
XM_017016970.1 | XP_016872459.1 | 14 | 3252 | |||
GenBank mRNA | AB007878.1 | - | 12 | 3035 | ||
AK056469.1 | - | 13 | 3008 | |||
AK294454.1 | - | 9 | 2825 | |||
AK294736.1 | - | 9 | 2863 | |||
AK295660.1 | - | 9 | 2929 | |||
AK302015.1 | - | 13 | 2792 | |||
AK304828.1 | - | 9 | 2841 | |||
AK315983.1 | - | 9 | 2868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
