siRNA Details
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015188.1 | NP_056003.1 | 3, 4 | 1393 | ||
XM_006717732.3 | XP_006717795.2 | 3, 4 | 1393 | |||
XM_006717733.2 | XP_006717796.1 | 4, 5 | 353 | |||
XM_011539558.2 | XP_011537860.1 | 3, 4 | 404 | |||
XM_017015996.1 | XP_016871485.1 | 3, 4 | 1393 | |||
XM_017015997.1 | XP_016871486.1 | 3, 4 | 1393 | |||
XM_017015998.1 | XP_016871487.1 | 2, 3 | 180 | |||
XM_017015999.1 | XP_016871488.1 | 2, 3 | 396 | |||
GenBank mRNA | AB011180.1 | - | 3, 4 | 1209 | ||
AB449898.1 | - | 3, 4 | 1190 | |||
AL049956.1 | - | 2, 3 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
