siRNA Details
Gene Symbol: ATG2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015104.2 | NP_055919.2 | 1, 2 | 288 | ||
XM_005273849.3 | XP_005273906.1 | 1, 2 | 279 | |||
XM_005273850.3 | XP_005273907.1 | 1, 2 | 279 | |||
XM_011544863.2 | XP_011543165.1 | 1, 2 | 279 | |||
XM_011544864.2 | XP_011543166.1 | 1, 2 | 279 | |||
XM_011544866.2 | XP_011543168.1 | 1, 2 | 279 | |||
XM_011544867.2 | XP_011543169.1 | 1, 2 | 279 | |||
GenBank mRNA | AB007864.1 | - | 1, 2 | 229 | ||
AK300925.1 | - | 1, 2 | 247 | |||
BC110650.1 | - | 1, 2 | 288 | 6147561 | ||
BC113090.1 | - | 1, 2 | 249 | |||
BC113091.1 | - | 1, 2 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
