siRNA Details
Gene Symbol: ATG2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015104.2 | NP_055919.2 | 41 | 6283 | ||
XM_005273849.3 | XP_005273906.1 | 41 | 6256 | |||
XM_005273850.3 | XP_005273907.1 | 41 | 6250 | |||
XM_011544863.2 | XP_011543165.1 | 41 | 6280 | |||
XM_011544864.2 | XP_011543166.1 | 41 | 6265 | |||
XM_011544865.2 | XP_011543167.1 | 41 | 6273 | |||
XM_011544868.2 | XP_011543170.1 | 26 | 3894 | |||
GenBank mRNA | AB007864.1 | - | 41 | 6224 | ||
BC008593.1 | - | 12 | 1911 | |||
BC027481.1 | - | 3 | 850 | |||
BC053596.1 | - | 16 | 2611 | |||
BC110650.1 | - | 41 | 6283 | 6147561 | ||
BC113090.1 | - | 10 | 1570 | |||
BC113091.1 | - | 7 | 1431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
