siRNA Details
Gene Symbol: PPM1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020700.1 | NP_065751.1 | 6 | 1207 | ||
XM_011538578.2 | XP_011536880.1 | 6 | 1343 | |||
XM_011538579.1 | XP_011536881.1 | 6 | 895 | |||
XM_017019676.1 | XP_016875165.1 | 6 | 1457 | |||
GenBank mRNA | AB032983.1 | - | 5 | 800 | ||
AB084258.1 | - | 6 | 1472 | |||
AK023315.1 | - | 7 | 1269 | |||
BC157843.1 | - | 6 | 1217 | 9057062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
