siRNA Details
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 9 | 1412 | ||
XM_011538078.2 | XP_011536380.1 | 9 | 1354 | |||
XM_017019083.1 | XP_016874572.1 | 9 | 1354 | |||
XM_017019084.1 | XP_016874573.1 | 10 | 1013 | |||
XM_017019085.1 | XP_016874574.1 | 11 | 1293 | |||
XM_017019086.1 | XP_016874575.1 | 11 | 1298 | |||
XM_017019087.1 | XP_016874576.1 | 9 | 1164 | |||
GenBank mRNA | AB023201.1 | - | 10 | 1007 | ||
AB449914.1 | - | 9 | 1104 | |||
AK055404.1 | - | 9 | 1354 | |||
AK299508.1 | - | 8 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
