siRNA Details
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015279.1 | NP_056094.1 | 12 | 2020 | ||
XM_011538078.2 | XP_011536380.1 | 13 | 2022 | |||
XM_017019083.1 | XP_016874572.1 | 12 | 1890 | |||
XM_017019084.1 | XP_016874573.1 | 14 | 1681 | |||
XM_017019085.1 | XP_016874574.1 | 14 | 1901 | |||
XM_017019086.1 | XP_016874575.1 | 14 | 1906 | |||
XM_017019087.1 | XP_016874576.1 | 12 | 1772 | |||
GenBank mRNA | AB023201.1 | - | 13 | 1615 | ||
AB449914.1 | - | 12 | 1712 | |||
AK055404.1 | - | 12 | 1962 | |||
AK299508.1 | - | 11 | 1799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
