siRNA Details
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257357.1 | NP_001244286.1 | 10, 11 | 2761 | ||
XM_011514600.2 | XP_011512902.1 | 10, 11 | 2858 | |||
XM_011514601.2 | XP_011512903.1 | 11, 12 | 2905 | |||
XM_011514602.2 | XP_011512904.1 | 10, 11 | 2825 | |||
XM_017010855.1 | XP_016866344.1 | 10, 11 | 4297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
