siRNA Details
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257357.1 | NP_001244286.1 | 10 | 2690 | ||
XM_011514600.2 | XP_011512902.1 | 10 | 2787 | |||
XM_011514601.2 | XP_011512903.1 | 11 | 2834 | |||
XM_011514602.2 | XP_011512904.1 | 10 | 2754 | |||
XM_011514604.2 | XP_011512906.1 | 10 | 2787 | |||
XM_017010855.1 | XP_016866344.1 | 10 | 4226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
