siRNA Details
Gene Symbol: TSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010873.2 | NP_001010873.1 | 2 | 442 | ||
NM_001159726.1 | NP_001153198.1 | 2 | 252 | |||
XM_011514396.2 | XP_011512698.1 | 2 | 245 | |||
XM_011514397.2 | XP_011512699.1 | 2 | 774 | |||
GenBank mRNA | BC104440.2 | - | 2 | 369 | ||
BC104441.2 | - | 2 | 369 | |||
BC137560.1 | - | 2 | 222 | 9021560 | ||
BC137565.1 | - | 2 | 222 | 9021565 | ||
BC144710.1 | - | 2 | 252 | 9053248 | ||
BU655765.1 | - | 2 | 452 | |||
BU657409.1 | - | 2 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
