siRNA Details
Gene Symbol: TBX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080508.2 | NP_001073977.1 | 3 | 609 | ||
XM_006715602.3 | XP_006715665.2 | 3 | 548 | |||
XM_011536247.2 | XP_011534549.2 | 3 | 548 | |||
XM_011536248.2 | XP_011534550.2 | 3 | 548 | |||
XM_011536249.2 | XP_011534551.2 | 3 | 548 | |||
XM_017011472.1 | XP_016866961.1 | 3 | 995 | |||
XM_017011473.1 | XP_016866962.1 | 3 | 548 | |||
GenBank mRNA | AJ010278.1 | - | 2 | 113 | ||
AK310179.1 | - | 3 | 287 | |||
BC040697.2 | - | 3 | 304 | |||
BC132715.1 | - | 3 | 609 | 40146737 | ||
BC157841.1 | - | 3 | 602 | 9057060 |

More Information
Other Species Targeted

Gene Ontology Categories:
