siRNA Details
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080433.1 | NP_001073902.1 | 6 | 2273 | ||
XM_005264125.1 | XP_005264182.1 | 5 | 1663 | |||
GenBank mRNA | AB067499.2 | - | 6 | 2360 | ||
BC131557.1 | - | 6 | 945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
