siRNA Details
Gene Symbol: GRIN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138690.2 | NP_619635.1 | 7 | 2502 | ||
XM_017026243.1 | XP_016881732.1 | 6 | 1748 | |||
GenBank mRNA | AY507106.1 | - | 7 | 2502 | ||
AY507107.1 | - | 7 | 2502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
