siRNA Details
Gene Symbol: FBXO42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018994.2 | NP_061867.1 | 2 | 491 | ||
XM_006710698.3 | XP_006710761.1 | 1 | 264 | |||
XM_017001502.1 | XP_016856991.1 | 2 | 454 | |||
XM_017001503.1 | XP_016856992.1 | 2 | 384 | |||
GenBank mRNA | AB037753.1 | - | 1 | 40 | ||
AK055598.1 | - | 2 | 454 | |||
AL833207.1 | - | 1 | 186 | |||
BC043410.1 | - | 2 | 456 | 6155685 | ||
BC063864.1 | - | 2 | 453 | 6182628 | ||
DA488838.1 | - | 2 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
