siRNA Details
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 2 | 984 | ||
XM_005271254.3 | XP_005271311.1 | 2 | 1662 | |||
XM_006710946.3 | XP_006711009.1 | 2 | 1663 | |||
XM_017002483.1 | XP_016857972.1 | 2 | 1664 | |||
XM_017002484.1 | XP_016857973.1 | 2 | 1663 | |||
XM_017002485.1 | XP_016857974.1 | 2 | 371 | |||
XM_017002486.1 | XP_016857975.1 | 2 | 284 | |||
GenBank mRNA | AL834286.1 | - | 2 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
