siRNA Details
Gene Symbol: DLGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080418.2 | NP_001073887.1 | 12 | 3607 | ||
XM_011541879.2 | XP_011540181.1 | 13 | 5492 | |||
XM_011541880.2 | XP_011540182.1 | 8 | 3316 | |||
GenBank mRNA | AF131778.1 | - | 5 | 1378 | ||
AK294907.1 | - | 4 | 951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
