siRNA Details
Gene Symbol: PRAMEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013407.2 | NP_001013425.2 | 3 | 591 |
Gene Symbol: PRAMEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146344.2 | NP_001139816.2 | 3 | 568 | ||
XM_011541479.2 | XP_011539781.1 | 2 | 754 | |||
GenBank mRNA | AL049680.1 | - | 3 | 567 | ||
BC144426.1 | - | 3 | 567 | 9052947 |
Gene Symbol: PRAMEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009611.4 | NP_001009611.2 | 3 | 568 | ||
GenBank mRNA | AL049681.1 | - | 3 | 568 |
Gene Symbol: PRAMEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010889.2 | NP_001010889.1 | 3 | 577 | ||
GenBank mRNA | BC101342.2 | - | 3 | 577 | 40025149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
