siRNA Details
Gene Symbol: PRAMEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013407.2 | NP_001013425.2 | 4 | 1112 | ||
GenBank mRNA | BC101344.1 | - | 4 | 1098 | 40025152 | |
BC130274.1 | - | 4 | 1097 | 40146305 | ||
BC130276.1 | - | 4 | 1097 | 40146307 |
Gene Symbol: PRAMEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146344.2 | NP_001139816.2 | 4 | 1089 | ||
XM_011541479.2 | XP_011539781.1 | 3 | 1275 | |||
GenBank mRNA | AL049680.1 | - | 4 | 1088 |
Gene Symbol: PRAMEF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310134.1 | NP_001297063.1 | 3 | 1034 |
Gene Symbol: PRAMEF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306072.1 | NP_001293001.1 | 3 | 1034 |
Gene Symbol: PRAMEF27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300891.1 | NP_001287820.1 | 3 | 1018 |
Gene Symbol: PRAMEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009611.4 | NP_001009611.2 | 4 | 1089 | ||
GenBank mRNA | AL049681.1 | - | 4 | 1089 |
Gene Symbol: PRAMEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010889.2 | NP_001010889.1 | 4 | 1098 | ||
GenBank mRNA | BC101342.2 | - | 4 | 1098 | 40025149 |
Gene Symbol: PRAMEF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010890.2 | NP_001010890.2 | 3 | 1018 | ||
GenBank mRNA | AK124292.1 | - | 4 | 1112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
