siRNA Details
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 14 | 2849 | ||
NM_001135181.1 | NP_001128653.1 | 15 | 2924 | |||
XM_011540924.2 | XP_011539226.1 | 14 | 4530 | |||
XM_011540926.2 | XP_011539228.1 | 13 | 3110 | |||
XM_011540927.1 | XP_011539229.1 | 12 | 2708 | |||
XM_011540928.1 | XP_011539230.1 | 14 | 2728 | |||
GenBank mRNA | AK131200.1 | - | 14 | 2802 | ||
BC025411.1 | - | 5 | 1649 | |||
BX648549.1 | - | 15 | 2878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
