siRNA Details
Gene Symbol: KIF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305792.1 | NP_001292721.1 | 1 | 672 | ||
NM_014875.2 | NP_055690.1 | 2 | 996 | |||
XM_011510231.2 | XP_011508533.1 | 3 | 1090 | |||
XM_011510232.2 | XP_011508534.1 | 2 | 752 | |||
XM_011510233.2 | XP_011508535.1 | 3 | 883 | |||
XM_011510235.2 | XP_011508537.1 | 3 | 595 | |||
XM_011510236.1 | XP_011508538.1 | 2 | 967 | |||
XM_017003005.1 | XP_016858494.1 | 1 | 1173 | |||
XM_017003006.1 | XP_016858495.1 | 2 | 781 | |||
XM_017003007.1 | XP_016858496.1 | 2 | 308 | |||
GenBank mRNA | BC113742.1 | - | 1 | 635 | 8322794 | |
D26361.1 | - | 2 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
