siRNA Details
Gene Symbol: ZC2HC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013623.2 | NP_001013645.1 | 4, 5 | 449 | ||
GenBank mRNA | BC146950.1 | - | 4, 5 | 449 | 9056878 | |
BC146969.1 | - | 4, 5 | 449 | 9056897 | ||
BC171862.1 | - | 5, 6 | 532 | 9054516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
