siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 52 | 8793 | ||
XM_006711956.2 | XP_006712019.1 | 52 | 8898 | |||
XM_011532649.2 | XP_011530951.1 | 52 | 8747 | |||
XM_011532650.2 | XP_011530952.1 | 52 | 8748 | |||
XM_011532652.1 | XP_011530954.1 | 51 | 8667 | |||
XM_011532653.1 | XP_011530955.1 | 51 | 8646 | |||
XM_011532654.1 | XP_011530956.1 | 51 | 8526 | |||
XM_011532655.2 | XP_011530957.1 | 48 | 7815 | |||
XM_011532657.1 | XP_011530959.1 | 46 | 7554 | |||
XM_017003521.1 | XP_016859010.1 | 50 | 8520 | |||
XM_017003522.1 | XP_016859011.1 | 47 | 8619 | |||
XM_017003523.1 | XP_016859012.1 | 47 | 7786 | |||
GenBank mRNA | AB051484.2 | - | 16 | 2728 | ||
AB290165.1 | - | 18 | 2932 | |||
BC117259.1 | - | 5 | 925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
