siRNA Details
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 56 | 9473 | ||
XM_006711956.2 | XP_006712019.1 | 56 | 9578 | |||
XM_011532649.2 | XP_011530951.1 | 56 | 9427 | |||
XM_011532650.2 | XP_011530952.1 | 56 | 9428 | |||
XM_011532652.1 | XP_011530954.1 | 55 | 9347 | |||
XM_011532653.1 | XP_011530955.1 | 55 | 9326 | |||
XM_011532654.1 | XP_011530956.1 | 55 | 9206 | |||
XM_011532655.2 | XP_011530957.1 | 52 | 8495 | |||
XM_011532657.1 | XP_011530959.1 | 50 | 8234 | |||
XM_017003521.1 | XP_016859010.1 | 54 | 9200 | |||
XM_017003522.1 | XP_016859011.1 | 51 | 9299 | |||
XM_017003523.1 | XP_016859012.1 | 51 | 8466 | |||
GenBank mRNA | AB051484.2 | - | 20 | 3408 | ||
AB290165.1 | - | 22 | 3612 | |||
BC117259.1 | - | 9 | 1605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
