siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014937.3 | NP_055752.1 | 5, 6 | 836 | ||
XM_006717720.3 | XP_006717783.1 | 5, 6 | 3223 | |||
XM_011539524.2 | XP_011537826.1 | 5, 6 | 599 | |||
XM_011539525.2 | XP_011537827.1 | 6, 7 | 3306 | |||
XM_011539527.2 | XP_011537829.2 | 4, 5 | 601 | |||
XM_011539528.2 | XP_011537830.1 | 4, 5 | 723 | |||
XM_017015935.1 | XP_016871424.1 | 5, 6 | 590 | |||
XM_017015936.1 | XP_016871425.1 | 8, 9 | 1224 | |||
GenBank mRNA | AB023183.1 | - | 5, 6 | 786 | ||
AK307758.1 | - | 5, 6 | 826 | |||
BC067820.1 | - | 5, 6 | 797 | |||
BC111493.1 | - | 5, 6 | 747 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
