siRNA Details
Gene Symbol: BTF3P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106717.2 | - | 1 | 36 | ||
BC106718.2 | - | 1 | 36 | |||
BC115358.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
