siRNA Details
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015258.1 | NP_056073.1 | 3, 4 | 373 | ||
XM_006717018.2 | XP_006717081.1 | 3, 4 | 290 | |||
XM_006717019.1 | XP_006717082.1 | 2, 3 | 221 | |||
XM_017014552.1 | XP_016870041.1 | 2, 3 | 221 | |||
GenBank mRNA | AB014574.1 | - | 3, 4 | 300 | ||
AK303074.1 | - | 2, 3 | 221 | |||
AK307507.1 | - | 7, 8 | 807 | |||
BC009609.1 | - | 3, 4 | 330 | |||
DC321217.1 | - | 3, 4 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
