siRNA Details
Gene Symbol: MLXIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014938.5 | NP_055753.3 | 17 | 5138 | ||
XM_006719290.3 | XP_006719353.1 | 18 | 5237 | |||
XM_006719291.3 | XP_006719354.1 | 18 | 5234 | |||
XM_006719292.3 | XP_006719355.1 | 17 | 5125 | |||
XM_006719294.3 | XP_006719357.1 | 12 | 4296 | |||
GenBank mRNA | AB020674.1 | - | 9 | 3985 | ||
AF245480.1 | - | 9 | 3985 | |||
AF245481.1 | - | 7 | 3161 | |||
BC017656.1 | - | 4 | 2676 | |||
BC151841.1 | - | 17 | 5120 | 8860321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
