siRNA Details
Gene Symbol: ATAD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242338.2 | NP_001229267.2 | 17 | 2600 | ||
NM_017552.3 | NP_060022.2 | 17 | 2600 | |||
NR_125717.1 | 17 | 2600 | ||||
XM_005264372.3 | XP_005264429.2 | 18 | 2642 | |||
XM_006712030.3 | XP_006712093.1 | 18 | 2642 | |||
XM_006712031.3 | XP_006712094.1 | 18 | 2642 | |||
XM_011532918.2 | XP_011531220.1 | 18 | 2642 | |||
XM_011532919.2 | XP_011531221.1 | 18 | 2642 | |||
XM_011532920.2 | XP_011531222.1 | 17 | 2600 | |||
XM_011532921.2 | XP_011531223.1 | 18 | 2642 | |||
XM_011532924.2 | XP_011531226.1 | 18 | 2642 | |||
XM_011532925.2 | XP_011531227.1 | 18 | 2642 | |||
XM_011532928.1 | XP_011531230.1 | 18 | 2642 | |||
XM_011532929.2 | XP_011531231.1 | 18 | 2642 | |||
XM_011532930.2 | XP_011531232.1 | 18 | 2642 | |||
XM_011532931.2 | XP_011531233.1 | 18 | 2642 | |||
XM_011532932.1 | XP_011531234.1 | 18 | 2642 | |||
XM_017004375.1 | XP_016859864.1 | 17 | 2600 | |||
GenBank mRNA | AB033066.1 | - | 1 | 98 | ||
AK125718.1 | - | 5 | 986 | |||
AK131301.1 | - | 5 | 740 | |||
BC171846.1 | - | 17 | 2547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
