siRNA Details
Gene Symbol: EQTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161585.1 | NP_001155057.1 | 1, 2 | 171 | ||
NM_020641.2 | NP_065692.2 | 1, 2 | 171 | |||
GenBank mRNA | AJ278482.1 | - | 1, 2 | 171 | ||
BC014307.1 | - | 1, 2 | 171 | |||
BC137351.1 | - | 1, 2 | 141 | 9021349 | ||
BC137352.1 | - | 1, 2 | 141 | 9021350 | ||
BC144602.1 | - | 1, 2 | 141 | 9053133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
