siRNA Details
Gene Symbol: TNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022093.1 | NP_071376.1 | 7 | 1626 | ||
XM_017002048.1 | XP_016857537.1 | 7 | 1631 | |||
XM_017002049.1 | XP_016857538.1 | 7 | 1632 | |||
GenBank mRNA | AK127044.1 | - | 7 | 1626 | ||
AL049689.1 | - | 6 | 1498 | |||
BC136619.1 | - | 6 | 1543 | 9020609 | ||
BC144305.1 | - | 6 | 1543 | 9052825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
