siRNA Details
Gene Symbol: TNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022093.1 | NP_071376.1 | 19 | 4564 | ||
XM_017002048.1 | XP_016857537.1 | 19 | 4569 | |||
XM_017002049.1 | XP_016857538.1 | 18 | 4306 | |||
GenBank mRNA | AK127044.1 | - | 19 | 4564 | ||
AK127654.1 | - | 9 | 1984 | |||
AL049689.1 | - | 18 | 4436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
