siRNA Details
Gene Symbol: SEPT7P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027269.1 | 3 | 458 | |||
GenBank mRNA | BC042387.1 | - | 3 | 458 | ||
BC048143.1 | - | 3 | 458 |
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 4 | 652 | |||
GenBank mRNA | AL832059.1 | - | 4 | 588 | ||
BC092503.1 | - | 4 | 597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
