siRNA Details
Gene Symbol: SPG20-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045180.1 | 3 | 228 | |||
NR_045181.1 | 2 | 141 | ||||
GenBank mRNA | AW292824.1 | - | 3 | 227 | ||
BF732820.1 | - | 2 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
