siRNA Details
Gene Symbol: CCDC169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144981.2 | NP_001138453.1 | 6 | 590 | ||
NM_001144982.2 | NP_001138454.1 | 5 | 510 | |||
NM_001144983.2 | NP_001138455.1 | 5 | 506 | |||
NM_001144984.2 | NP_001138456.1 | 4 | 469 | |||
NM_001144985.2 | NP_001138457.1 | 5 | 510 | |||
NM_001144986.2 | NP_001138458.1 | 5 | 506 | |||
NM_001198908.1 | NP_001185837.1 | 6 | 590 | |||
GenBank mRNA | AK308999.1 | - | 5 | 479 | ||
BC037403.1 | - | 6 | 721 | |||
BC146975.1 | - | 6 | 484 | 9056903 | ||
BC171867.1 | - | 5 | 401 | 9054521 | ||
BC171872.1 | - | 5 | 404 | 9054526 | ||
BC171874.1 | - | 4 | 363 | 9054528 | ||
BC171880.1 | - | 5 | 400 | 9054534 | ||
BC171881.1 | - | 4 | 363 | 9054535 | ||
BC171882.1 | - | 5 | 400 | 9054536 | ||
BC171884.1 | - | 5 | 404 | |||
DN999189.1 | - | 6 | 547 |
Gene Symbol: CCDC169-SOHLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198910.1 | NP_001185839.1 | 4 | 469 | ||
GenBank mRNA | AK301863.1 | - | 4 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
